missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptogyrin 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77102-0.025mg
This item is not returnable.
View return policy
Description
Synaptogyrin 3 Polyclonal specifically detects Synaptogyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Synaptogyrin 3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| MGC20003, synaptogyrin 3, synaptogyrin-3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| NP_004200 | |
| SYNGR3 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human SYNGR3. The immunogen is located within the last 50 amino acids of SYNGR3. Amino Acid Squence: ETYQSPFFTETLDTSPKGY | |
| 0.025 mg | |
| Stem Cell Markers | |
| 9143 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction