missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Synaptogyrin 2 Polyclonal antibody specifically detects Synaptogyrin 2 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Synaptogyrin 2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | cellugyrin, MGC102914, synaptogyrin 2, synaptogyrin-2 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human SYNGR2. The immunogen is located within amino acids 30 - 80 of SYNGR2. Amino Acid Squence: IYGEGYSNAHESKQMY |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?