missing translation for 'onlineSavingsMsg'
Learn More

Synapsin II Rabbit anti-Human, Mouse, Rat, Clone: 7J6L0, Novus Biologicals™

Product Code. 18309644
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18309644 20 μg 20µL
18382842 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18309644 Supplier Novus Biologicals Supplier No. NBP31576520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Synapsin II Monoclonal antibody specifically detects Synapsin II in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synapsin II
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 7J6L0
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human Synapsin II (Q92777).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6854
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.