missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synapsin II Monoclonal Antibody (GT1446)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547281
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.
Specifications
| Synapsin II | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 20% glycerol and no preservative | |
| Q63537, Q64332, Q92777 | |
| SYN2 | |
| Recombinant protein encompassing a sequence within the center region of human Synapsin II. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT1446 | |
| Unconjugated | |
| SYN2 | |
| 2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb | |
| Mouse | |
| Protein A | |
| RUO | |
| 20965, 29179, 6854 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction