missing translation for 'onlineSavingsMsg'
Learn More

Synapsin II Antibody (GT965), Novus Biologicals™

Product Code. 18391143
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391143 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391143 Supplier Bio-Techne Supplier No. NBP313690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Synapsin II Monoclonal antibody specifically detects Synapsin II in Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synapsin II
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone GT965
Conjugate Unconjugated
Formulation PBS, 20% Glycerol
Gene Alias synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human Synapsin II II. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6854
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.