missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synapsin I, p Ser9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-180
This item is not returnable.
View return policy
Description
Synapsin I Polyclonal specifically detects Synapsin I in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Synapsin I (p Ser9) | |
| Polyclonal | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| phospho Synapsin, SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI | |
| Rabbit | |
| 78 kDa | |
| 0.1 mL | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 6853 | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 0.1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:500 | |
| P09951 | |
| SYN1 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser9 conjugated to KLH. Accession # P17599 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for ∼78k synapsin I doublet protein phosphorylated at Ser9. The antibody also weakly labels the ∼55k synapsin II protein which has a similar phosphorylation site to that of Ser9 on synapsin I. Immunolabeling is blocked by preadsorption of the antibody with the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. Immunolabeling is also completely eliminated by treatment with lambda phosphatase. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction