missing translation for 'onlineSavingsMsg'
Learn More

syn-2-Pyridinealdoxime, 99+%

Product Code. 10644992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10644992 100 g Glass bottle 100g
10458290 25 g Glass bottle 25g
10783275 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10644992 Supplier Thermo Scientific Acros Supplier No. 131791000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1193-96-0 | C6H6N2O | 122.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-96-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00167166
InChI Key MTFJSAGADRTKCI-UHFFFAOYSA-N
Synonym pyridine-2-aldoxime, picolinaldehyde oxime, 2-pyridinecarbaldehyde oxime, z-pyridine-2-carbaldehyde oxime, 2-nitrosomethylidene-1h-pyridine, 2-pyridinealdoxime, e-picolinaldehyde oxime, 2-hydroxyiminomethyl-pyridine, 2-pyridine aldoxime, 2-pyridinecarboxaldehyde, oxime, e
PubChem CID 6740772
IUPAC Name 2-(nitrosomethylidene)-1H-pyridine
SMILES ON=CC1=CC=CC=N1

Specifications

Melting Point 110°C to 113°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 g
Beilstein 21, I, 288
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: may decompose,265g/L ethanol (20°C),31% in methanol- 18% in acetone (20°C),0.5% in toluene- 0.5% in gasoline (20°C)
Formula Weight 122.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material syn-2-Pyridinealdoxime
Show More Show Less
compliance-icons
Product Identifier
  • syn-2-Pyridinealdoxime
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.