missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation Symmetry C4 Method Validation Kit

Product Code. p-7236844
Encompass
Click to view available options
Length (Metric):
150 mm
250 mm
Particle Size:
3.5 μm
5 μm
Unit Size:
Pack of 3
This item is not returnable. View return policy

Product Code. 13242510

Brand: Waters Corporation 186000291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods.

  • No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
  • Available in a variety of particle sizes, chemistries and dimensions.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 300 Å
Quantity 3/Pk.
USP Type L26
Endcapped Yes
Type Method Validation Kit
For Use With (Application) Protein
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.