missing translation for 'onlineSavingsMsg'
Learn More

SYM2206, MedChemExpress

Product Code. 16490058
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490058

Brand: Medchem Express HY18689

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SYM2206 is a potent and non-competitive AMPA receptor antagonist, with an IC50 of 1.6 μM. SYM2206 blocks Nav1.6-mediated persistent currents.

SYM2206 is a potent and non-competitive AMPA receptor antagonist, with an IC50 of 1.6 μM. SYM2206 blocks Nav1.6-mediated persistent currents.

Specifications

Chemical Name or Material SYM2206
CAS 173952-44-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H22N4O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (272.92 mM; Need ultrasonic)
SMILES O=C(N1N=C(C2=CC=C(N)C=C2)C3=C(C=C4C(OCO4)=C3)C1C)NCCC
Molecular Weight (g/mol) 366.41
Formula Weight 366.41
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.