missing translation for 'onlineSavingsMsg'
Learn More

sym-Diphenylcarbazide, 98%, ACS reagent

Product Code. 10437000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10437000 100 g Glass bottle 100g
10427000 25 g Glass bottle 25g
11925661 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10437000 Supplier Thermo Scientific Acros Supplier No. 158841000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-22-7 | C13H14N4O | 242.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.28
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

Melting Point 173.0°C to 176.0°C
Color Beige to Pink
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 15,292
Sensitivity (to chromate) passes test
Merck Index 15,3352
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and acetone
Formula Weight 242.28
Percent Purity 98%
Grade ACS Reagent
Physical Form Powder or Flakes
Chemical Name or Material sym-Diphenylcarbazide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.