missing translation for 'onlineSavingsMsg'
Learn More

sym-Diformylhydrazine, 97%, Thermo Scientific Chemicals

Product Code. 10233950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10233950 25 g Glass bottle 25g
10479020 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10233950 Supplier Thermo Scientific Acros Supplier No. 114640250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-36-4
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.07
MDL Number MFCD00003275
InChI Key POVXOWVFLAAVBH-UHFFFAOYSA-N
Synonym 1,2-diformylhydrazine, n'-formylformohydrazide, diformylhydrazine, 1,2-hydrazinedicarboxaldehyde, bicarbamaldehyde, diformohydrazide, s-diformohydrazide, s-diformylhydrazine, n,n'-diformylhydrazine, n'-formylformic hydrazide
PubChem CID 12342
SMILES O=CNNC=O

Specifications

Melting Point 155°C to 160°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HCONHNHCHO
Quantity 25 g
Beilstein 02, 93
IUPAC Name N-formamidoformamide
Formula Weight 88.06
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material sym-Diformylhydrazine, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.