missing translation for 'onlineSavingsMsg'
Learn More

SYM 2081, MedChemExpress

Product Code. 16456057
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456057 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16456057 Supplier Medchem Express Supplier No. HY101310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SYM 2081 is a high-affinity ligand and potent, selective agonist of kainate receptors, inhibits [3H]-kainate binding with an IC50 of 35 nM, almost 3000- and 200-fold selectivity for kainate receptors over AMPA and NMDA receptors respectively.

SYM 2081 is a high-affinity ligand and potent, selective agonist of kainate receptors, inhibits [3H]-kainate binding with an IC50 of 35 nM, almost 3000- and 200-fold selectivity for kainate receptors over AMPA and NMDA receptors respectively.

Specifications

Chemical Name or Material SYM 2081
Physical Form Powder
CAS 31137-74-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C6H11NO4
For Use With (Application) Neuroscience-Neurodegeneration
SMILES N[C@@H](C[C@@H](C)C(O)=O)C(O)=O
Molecular Weight (g/mol) 161.16
Formula Weight 161.16
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.