missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syk Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528574
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, Raji. Predicted reactivity: Mouse (89%), Rat (89%), Pig (90%), Chicken (81%), Rhesus Monkey (91%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Spleen tyrosine kinase (SYK), along with ZAP70, are cytoplasmic tyrosine kinases that are viewed as potential druge targets for allergic disorders and autoimmune diseases. SYK may also play a role in cancer. Syk translocates to the plasma membrane upon B cell antigen receptor (BCR) or the high-affinity IgE receptor (FcepsilonRI) triggering, and phosphorylates downstream adaptor proteins, thereby providing docking sites for initiation of subsequent signaling pathways, such as calcium mobilization, cytoskeleton remodeling, or transcription of specific genes. Syk binds to the receptor assemblies through interactions of its pair of SH2 domains with ITAM motives of the receptor, which have been phosphorylated by Src-family kinases. These kinases also help to activate Syk by phosphorylation of its activation loop.
Specifications
| Syk | |
| Polyclonal | |
| Unconjugated | |
| Syk | |
| DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20963, 25155, 6850 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.02 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P43405, P48025, Q64725 | |
| Syk | |
| Recombinant protein encompassing a sequence within the center region of human Syk. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction