missing translation for 'onlineSavingsMsg'
Learn More

SYK Mouse anti-Human, Clone: 4C4A12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66721-1-IG-20UL

 View more versions of this product

Product Code. 16848865

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the family of non-receptor type Tyr protein kinases. This protein is widely expressed in hematopoietic cells and is involved in coupling activated immunoreceptors to downstream signaling events that mediate diverse cellular responses, including proliferation, differentiation, and phagocytosis. It is thought to be a modulator of epithelial cell growth and a potential tumor suppressor in human breast carcinomas. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SYK
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P43405
SYK
SYK Fusion Protein Ag17654
20 μL
Primary
Human
Antibody
IgG2b
Western Blot
4C4A12
Unconjugated
SYK
DKFZp313N1010, FLJ25043, FLJ37489, spleen tyrosine kinase, SYK, Tyrosine protein kinase SYK
Mouse
Protein A
RUO
6850
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.