missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYF2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16958-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| SYF2 | |
| Polyclonal | |
| Unconjugated | |
| SYF2 | |
| CBPIN, CCNDBP1 interactor, fSAP29, GCIPIP, NTC31, P29, Pre mRNA splicing factor SYF2, SYF2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25949 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95926 | |
| SYF2 | |
| SYF2 Fusion Protein Ag10656 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction