Learn More
Invitrogen™ SYF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110453
Description
Immunogen sequence: MAAIAASEVL VDSAEEGSLA AAAELAAQKR EQRLRKFREL HLMRNEARKL NHQEVVEEDK RLKLPANWEA KKARLEWELK EEEKKKECAA RGEDYEKVKL LEISAEDAER WERKKKRKNP DLGFSDYAAA QLRQYHRLTK QIKPDMETYE RLREKHGEEF FPTSNSLLHG THVPSTEEID RMVIDLEKQI EKRDKYSRRR PYNDDADIDY INERNAKFNK KAERFYGKYT AEIKQNLERG TAV.
This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| SYF2 | |
| Polyclonal | |
| Unconjugated | |
| SYF2 | |
| 1110018L13Rik; Cbpin; CCNDBP1 interactor; CCNDBP1-interactor; D4Bwg1551e; DKFZP564O2082; fSAP29; functional spliceosome-associated protein 29; GCIP-interacting protein p29; GCIP-interacting protein p29 (Gcipip); Gcipip; mp29; Ntc31; P29; Pre-mRNA-splicing factor SYF2; SYF2; SYF2 homolog, RNA splicing factor; SYF2 homolog, RNA splicing factor (S. cerevisiae); SYF2 pre-mRNA splicing factor; SYF2 pre-mRNA-splicing factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 68592 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9D198 | |
| SYF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-243 of human SYF2 (NP_056299.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.