missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYDE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89350
This item is not returnable.
View return policy
Description
SYDE1 Polyclonal specifically detects SYDE1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SYDE1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Q6ZW31 | |
| SYDE1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GDWSVCGRDFLPCGRDFLSGPDYDHVTGSDSEDEDEEVGEPRVTGDFEDDFDAPFNPHLNLKDFDALILDLERELSKQINV | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 2.4.1.25, EC 2.6.1.9, FLJ13511, Protein syd-1 homolog 1, rho GTPase-activating protein SYDE1, synapse defective 1, Rho GTPase, homolog 1 (C. elegans), Synapse defective protein 1 homolog 1,7h3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 85360 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction