missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYCE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11063-1-AP-150UL
This item is not returnable.
View return policy
Description
SYCE1 (synaptonemal complex central element protein 1), also known as CT76 (cancer/testis antigen 76), is a 351 amino acid nuclear protein that belongs to the SYCE family and exists as three alternatively spliced isoforms. Colocalizing with SYCE2 on chromatin, SYCE1 exists as a homodimer and is found in the transverse central elements of the central region between lateral elements of the synaptonemal complexes (SCS). The transverse central element of the SCS is formed between homologous chromosomes during meiotic prophase and requires SYCP1.Specifications
| SYCE1 | |
| Polyclonal | |
| Unconjugated | |
| SYCE1 | |
| C10orf94, Cancer/testis antigen 76, CT76, SYCE1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 502372, 74075, 93426 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5XHZ2, Q8N0S2, Q9D495 | |
| Syce1 | |
| SYCE1 Fusion Protein Ag1527 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction