Learn More
Invitrogen™ SYBR GreenER™ qPCR SuperMix for ABI PRISM™ Instrument
Description
The SYBR™ GreenER™ qPCR SuperMix kits include a dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.
Consistent Specificity and Low-Copy Detection: The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.
- The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility
- A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
- An optimized buffer system - improves sensitivity and provides excellent long-term stability
- Uracil DNA glycosylase - reduces carryover contamination in qPCR
Order Info
Shipping Conditions: Dry Ice
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Contains SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ (12.5 mL). SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ is a 2X SuperMix with premixed ROX Reference dye. Sufficient reagents are provided for 500 reactions based on a 50 μL reaction size. Store at +4°C upon receipt. |
| Detection Method | SYBR |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System |
| Product Line | Platinum, SYBR GreenER |
| Product Type | Real Time PCR SYBR Master Mix |
| Quantity | 500 reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.