missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress SYBR Green qPCR Master Mix (Low ROX)
Specifications
Specifications
| Content And Storage | Stored at -20°C protecting from light, and is stable for up to 18 months. Avoid repetitive freeze-thaw cycles while using. For immediate use, components may be stored at 2°C to 8°C. |
| Product Type | Kits |
| Quantity | 2000 Reactions |
| Shipping Condition | Room temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?