Learn More
MedChemExpress SYBR Green qPCR Master Mix
Product Code. 16492326
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 Reactions
Unit Size:
500 reactions
Product Code. Quantity Unit Size
16492326 500 Reactions 500 reactions
1 options
Product Code. 16492326 Supplier MedChemExpress Supplier No. HYK0501
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MedChemExpress MCE qPCR Master Mix is provided as a simple-to-use, stabilized 2X formulation that includes all components for qPCR except sample DNA, primers and water.
Specifications
Content And Storage Stored at -20°C protecting from light, and is stable for up to 18 months. Avoid repetitive freeze-thaw cycles while using. For immediate use, components may be stored at 2°C to 8°C.
Product Type Kits
Quantity 500 Reactions
Shipping Condition Room temperature