missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR Green I Nucleic Acid Gel Stain, MedChemExpress
Click to view available options
Quantity:
100 μL
500 μL
Unit Size:
100µL
500µL
Description
SYBR Green I Nucleic Acid Gel Stain is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels.
Specifications
Specifications
| Chemical Name or Material | SYBR Green I Nucleic Acid Gel Stain |
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 500 μL |
| Purity Grade Notes | Research |
| Grade | Research |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction