missing translation for 'onlineSavingsMsg'
Learn More

SYBR Green I Nucleic Acid Gel Stain, MedChemExpress

Product Code. 16458946
Encompass_Preferred
Click to view available options
Quantity:
100 μL
500 μL
Unit Size:
100µL
500µL
This item is not returnable. View return policy

Product Code. 16458946

Brand: Medchem Express HYK1004

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress SYBR Green I Nucleic Acid Gel Stain is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels.

SYBR Green I Nucleic Acid Gel Stain is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels.

Specifications

Chemical Name or Material SYBR Green I Nucleic Acid Gel Stain
Physical Form Liquid
Color Colorless
Quantity 100 μL
Purity Grade Notes Research
Grade Research

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.