Learn More
SymLine SwitchBox
Click to view available options
:
Electronic Switchbox
Description
Improved safety through increased collecting volume, the reserve canister protects against overflowing - even for a longer time period. For example, waste can be disposed of over the weekend without operator supervision and without having to empty or change the full container.
- Switchbox compatible to all electronic SCAT and SymLine level sensors as well as the Signalbox T5 and Built-in Signalbox2
- Established and professional disposal solution for laboratories with increased disposal needs
- Many more applications, not only for waste disposal of HPLC systems
Specifications
Specifications
| For Use With (Application) | Automatically Divert Waste Liquids Into The Reserve Canister. |
| For Use With (Equipment) | 3-Way Ball Valve |
| Type | Control Unit |