missing translation for 'onlineSavingsMsg'
Learn More

Swine Serum Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-44106

Product Code. 18128687

  • £388.00 / 2mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifically detects Swine Serum in Porcine samples, and it is validated for ELISA, Immunoprecipitation, Western Blotting

Swine Serum Polyclonal specifically detects Swine Serum in Porcine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Swine Serum
Polyclonal
Unconjugated
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Rabbit
Delipidation and Defibrination
RUO
Reconstitute with 2.0 ml deionized water (or equivalent)
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunoprecipitation
LYOPH
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Swine Sera
Swine Serum Proteins
2 mL
Primary
Pig
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only