Learn More
Thermo Scientific™ Spring Loaded Transfer Line Nut for ISQ and TSQ
Shop All Thermo Scientific ProductsDescription
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Specifications
Specifications
| Description | Chromatography Accessories |
| For Use With (Equipment) | Swap-IT |