Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories
Product Code. 10437704 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Ferrule
SWAP-Tight Ferrule 0.1 to 0.25 mm
SWAP-Tight Ferrule 0.32 mm
SWAP-Tight Ferrule, Blank
SWAP-Tight Nut
SWAP-Tight Nut and Adaptor
Transfer Line Nut
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10437704 SWAP-Tight Ferrule, Blank Pack of 10
10331055 Transfer Line Nut Each
10351555 SWAP-Tight Nut Each
10117414 Transfer Line Nut Each
10085644 Ferrule Pack of 10
10279264 SWAP-Tight Ferrule 0.1 to 0.25 mm Pack of 10
10249794 SWAP-Tight Ferrule 0.32 mm Pack of 10
10249504 SWAP-Tight Nut and Adaptor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10437704 Supplier Thermo Scientific™ Supplier No. 290SG000
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
TRUSTED_SUSTAINABILITY
Specifications
Type SWAP-Tight Ferrule, Blank
Quantity 10 Pk.
For Use With (Equipment) SWAP-IT GC-MS Interface System
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.