Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
£385.00 - £653.00
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | For Use With (Equipment) | Includes | Preis | Menge & Verfügbarkeit | ||||
15237508
|
Thermo Scientific™
60180-791 |
ISQ GC-MS, TSQ 8000 GC-MS/MS | Interface, Nuts, Ferrules, and Accessories |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10702327
|
Thermo Scientific™
60180-794 |
Agilent GC-MS Instruments | Interface, Nuts, Ferrules, and Accessories |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10230435
|
Thermo Scientific™
60180-790 |
DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only | Interface, Nuts, Ferrules, and Accessories |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10075924
|
Thermo Scientific™
60180-792 |
DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only | SWAP-IT Interface only |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10270005
|
Thermo Scientific™
60180-796 |
Agilent GC-MS Instruments | SWAP-IT Interface only |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15247508
|
Thermo Scientific™
60180-793 |
ISQ GC-MS, TSQ 8000 GC-MS Systems Only | SWAP-IT Interface only |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Beschreibung
Make column changeovers quick and easy. The Thermo Scientific™ SWAP-IT™ GC-MS Interface system is a powerful tool that enables capillary column changes without the need to vent your mass spectrometer (MS). It uses a specially deactivated, robust stainless steel transfer capillary as an interface between the analytical column and the MS detector. The small inner diameter of transfer capillary ensures minimum exposure to air and water when the analytical column is disconnected. A helium purge is not required. Finger-tight connections make it quick and easy without the need for tools.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.