Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System
Product Code. 15247508 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Agilent GC-MS Instruments
DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only
ISQ GC-MS, TSQ 8000 GC-MS Systems Only
ISQ GC-MS, TSQ 8000 GC-MS/MS
Includes:
Interface, Nuts, Ferrules, and Accessories
SWAP-IT Interface only
Unit Size:
Each
Product Code. 15247508
Brand: Thermo Scientific™ 60180793
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Make column changeovers quick and easy. The Thermo Scientific™ SWAP-IT™ GC-MS Interface system is a powerful tool that enables capillary column changes without the need to vent your mass spectrometer (MS). It uses a specially deactivated, robust stainless steel transfer capillary as an interface between the analytical column and the MS detector. The small inner diameter of transfer capillary ensures minimum exposure to air and water when the analytical column is disconnected. A helium purge is not required. Finger-tight connections make it quick and easy without the need for tools.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Spezifikation
Type GC-MS Interface System
Description SWAP-IT GC-MS Interface Unit
Includes SWAP-IT Interface only
For Use With (Equipment) ISQ GC-MS, TSQ 8000 GC-MS Systems Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt