Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System
Product Code. 15237508 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Agilent GC-MS Instruments
DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only
ISQ GC-MS, TSQ 8000 GC-MS Systems Only
ISQ GC-MS, TSQ 8000 GC-MS/MS
Includes:
Interface, Nuts, Ferrules, and Accessories
SWAP-IT Interface only
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Includes unitSize
15237508 ISQ GC-MS, TSQ 8000 GC-MS/MS Interface, Nuts, Ferrules, and Accessories Each
10702327 Agilent GC-MS Instruments Interface, Nuts, Ferrules, and Accessories Each
10230435 DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only Interface, Nuts, Ferrules, and Accessories Each
10075924 DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only SWAP-IT Interface only Each
10270005 Agilent GC-MS Instruments SWAP-IT Interface only Each
15247508 ISQ GC-MS, TSQ 8000 GC-MS Systems Only SWAP-IT Interface only Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15237508 Supplier Thermo Scientific™ Supplier No. 60180791
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Make column changeovers quick and easy. The Thermo Scientific™ SWAP-IT™ GC-MS Interface system is a powerful tool that enables capillary column changes without the need to vent your mass spectrometer (MS). It uses a specially deactivated, robust stainless steel transfer capillary as an interface between the analytical column and the MS detector. The small inner diameter of transfer capillary ensures minimum exposure to air and water when the analytical column is disconnected. A helium purge is not required. Finger-tight connections make it quick and easy without the need for tools.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type GC-MS Interface Kit
Description SWAP-IT GC-MS Interface Kit
Includes Interface, Nuts, Ferrules, and Accessories
For Use With (Equipment) ISQ GC-MS, TSQ 8000 GC-MS/MS