Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Brand: Thermo Scientific™ 60180-796
Code : 16
Additional Details : Weight : 0.50000kg
Description
Make column changeovers quick and easy. The Thermo Scientific™ SWAP-IT™ GC-MS Interface system is a powerful tool that enables capillary column changes without the need to vent your mass spectrometer (MS). It uses a specially deactivated, robust stainless steel transfer capillary as an interface between the analytical column and the MS detector. The small inner diameter of transfer capillary ensures minimum exposure to air and water when the analytical column is disconnected. A helium purge is not required. Finger-tight connections make it quick and easy without the need for tools.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
3-Port SilFlow MCD | |
SWAP-IT GC-MS Interface Unit | |
SWAP-IT Interface only | |
Agilent GC-MS Instruments |