missing translation for 'onlineSavingsMsg'
Learn More

LTE Scientific™ Swallow Oven, 480 L, Multi-Program with Fan Circulation

Product Code. 15583507
Click to view available options
Capacity (Metric):
480 L
This item is not returnable. View return policy

Product Code. 15583507

Brand: LTE Scientific™ LO/SWL/20/1

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Swallow Oven comes with 3 shelves.

  • Swallow Series

Specifications

Capacity (Metric) 480 L
Temperature (Metric) 40°C to 250°C
Voltage 220/240 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.