Learn More
Neogen Swab-Sampler
Product Code. 17258520
Encompass
Change view
Click to view available options
For Use With (Application):
1 mL Letheen Broth
4 mL Letheen Broth
4 mL Neutralizing Buffer
5 mL Letheen Broth
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
17258520 5 mL Letheen Broth Pack of 100
17228520 1 mL Letheen Broth Pack of 100
17238520 4 mL Letheen Broth Pack of 100
17248520 4 mL Neutralizing Buffer Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17258520 Supplier Neogen Supplier No. 7000030302
Convenient and quick collection device for environmental sampling and testing over small surface areas.
  • Letheen Broth vial is ready to use for pathogen testing of environmental samples.
  • Swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Bottles have ample headspace for sample addition and vortexing.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifications
For Use With (Application) 5 mL Letheen Broth
Quantity 100 /Cs.
Product Type Swab