Learn More
Neogen Swab-Sampler
Description
- Letheen Broth vial is ready to use for pathogen testing of environmental samples.
- Swab reaches into corners and hard-to-reach areas for increased accuracy.
- Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
- Bottles have ample headspace for sample addition and vortexing.
- Self-contained swab and vial for increased productivity and efficiency.
Specifications
Specifications
| For Use With (Application) | 1 mL Letheen Broth |
| Quantity | 100 /Cs. |
| Product Type | Swab |