missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SW033291, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
SW033291 is a potent and high-affinity inhibitor of 15-PGDH with a Ki of 0.1 nM. SW033291 increases prostaglandin PGE2 levels in bone marrow and other tissues. SW033291 also promotes tissue regeneration.
Specifications
Specifications
| Chemical Name or Material | SW033291 |
| CAS | 459147-39-8 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20N2OS3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 33.33 mg/mL (80.78 mM; ultrasonic and warming and heat to 70°C) ∣Ethanol : 25 mg/mL (60.59 mM; Need ultrasonic) |
| SMILES | NC1=C(S(CCCC)=O)SC2=NC(C3=CC=CS3)=CC(C4=CC=CC=C4)=C21 |
| Molecular Weight (g/mol) | 412.59 |
| Formula Weight | 412.59 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction