missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SVOPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569936
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 93%; Zebrafish: 92%.
SVOPL (putative transporter SVOPL), also known as SV2-related protein-like, is a 492 amino acid multi-pass membrane protein belonging to the major facilitator superfamily. SVOPL is a paralog to synaptic vesicle protein (SVOP) that exists as two alternatively spliced isoforms. The gene encoding SVOPL maps to human chromosome 7q34. Chromosome 7 is approximately 158 milllion bases long, encodes over 1,000 genes and makes up approximately 5% of the human genome. Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. Deletions of portions of the q arm of chromosome 7 are linked to myeloid disorders, including acute myelogenous leukemia and myelodysplasia.
Specifications
| SVOPL | |
| Polyclonal | |
| Unconjugated | |
| SVOPL | |
| 9430071P14Rik; LOC518832 protein; putative transporter SVOPL; RGD1566053; si:dkeyp-27e10.2; similar to RIKEN cDNA 9430071P14 gene; SV2 related protein homolog (rat)-like; SV2 related protein homolog-like; SV2-related protein-like; SVOP like; Svopl; SVOP-like; SVOP-like protein; zgc:101741 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 136306 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N434 | |
| SVOPL | |
| synthetic peptide directed towards the C-terminal region of human SVOPL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction