missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SVOC Mixture 231 100 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Specifications
Specifications
| CAS | 4901-51-3, 58-90-2, 15950-66-0, 935-95-5, 933-78-8, 697-82-5 |
| Quantity | 10 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction