missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SVL™ Torion Spare Fitting Joints
Description
- Glass filled polypropylene for use at temperatures up to 180°C
- PTFE for use at temperatures up to 250°C
Specifications
Specifications
| Type | Polypropylene Clamp |
| Thickness (Metric) Wall | 8.7mm (Bore) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?