missing translation for 'onlineSavingsMsg'
Learn More

SVL™ Torion Spare Fitting Joints

Product Code. 12563346
Change view
Click to view available options
Thickness (Metric) Wall:
14.7mm (Bore)
5.7mm (Bore)
8.7mm (Bore)
Type:
PTFE Clamp
Polypropylene Clamp
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Wall Type unitSize
12563346 14.7mm (Bore) Polypropylene Clamp Pack of 10
12698405 8.7mm (Bore) PTFE Clamp Pack of 10
15312969 5.7mm (Bore) Polypropylene Clamp Pack of 10
12493531 14.7mm (Bore) PTFE Clamp Pack of 10
13431819 8.7mm (Bore) Polypropylene Clamp Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12563346 Supplier SVL Supplier No. 72003
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Torion glass bodies and rubber/PTFE sealing ring

  • Glass filled polypropylene for use at temperatures up to 180°C
  • PTFE for use at temperatures up to 250°C
TRUSTED_SUSTAINABILITY

Specifications

Type Polypropylene Clamp
Thickness (Metric) Wall 14.7mm (Bore)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.