Learn More
SVL™ Coupling for Butt Joint
Product Code. 12073123
Click to view available options
Size:
Joint Size: 15
Joint Size: 22
Joint Size: 42
Unit Size:
Each
Pack of 10
Product Code. 12073123
Brand: SVL 70111
Only null left
Add to basket
Add to basket
Plastic coupling and sealing rings designed to secure two SVR™ glass screwthreads of the same dia. In a butt joint
  • Plastic coupling and sealing ring designed to secure two SVL™ glass screwthreads of the same diameter in a butt joint
  • Coupling is manufactured from phenolic plastic and consists of two separately threaded sections
  • Sealing ring is manufactured from silicone rubber and sheathed with PTFE for protection against chemical attack. Internal diameter of the ring corresponds to the bore of the SVL™ glass screwthread so that flow is not restricted
  • Heat resistant up to 200°C
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SVL and Quickfit Glassware
For Use With (Application) To secure two SVL Glass screw threads of the same diameter in a butt joint
Includes Sealing ring
Quantity 10 Pk.
Size Joint Size: 15
Type SVL coupling