Learn More
Quickfit™ Claisen Still Head Adapter
Product Code. 12048493
Change view
Click to view available options
Joint Type:
Condenser: 19/26, Flask: 24/29
Condenser: 24/29, Flask: 24/29
Size:
Socket: 14/23
Unit Size:
Each
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Type Size unitSize
12048493 Condenser: 24/29, Flask: 24/29 Socket: 14/23 Each
12038493 Condenser: 19/26, Flask: 24/29 Socket: 14/23 Each
15537970 - - Pack of 5
15547970 - - Pack of 5
15557970 - - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12048493 Supplier Quickfit Supplier No. SH6/33
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PTFE joint sleeves eliminate the possibility of joint seizure
  • Similar to the SH4: still heads but with extra socket to accept dropping funnel or stirrer
TRUSTED_SUSTAINABILITY
Specifications
Type Claisen Still Head
Material Borosilicate Glass
Standard Taper Joint 24/49 (Condenser), 24/29 (Flask)
Joint Type Condenser: 24/29, Flask: 24/29
Size Socket: 14/23