Learn More
SVL™ Borosilicate Glass Lid for SVL™ Reaction Vessel
Product Code. 15577690
Change view
Click to view available options
Height (Metric):
100 mm
Unit Size:
Pack of 5
Product Code. Height (Metric) unitSize
15577690 100 mm Pack of 5
1 options
Product Code. 15577690 Supplier SVL Supplier No. 29602
Add to basket
Add to basket
For use with SVL™ reaction vessels
  • For use with SVL™ reaction vessels
  • Borosilicate glass construction available in three styles: - Plain with knob (no necks) to act as a cover - 296-00 - With two necks (one vertical central neck and one vertical side neck) - With five necks as shown in the table below
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
For Use With (Equipment) SVL™ reaction vessels
Height (Metric) 100 mm
Product Type Lid