Learn More
SV40 Small T antigen Antibody (pab 280) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-61248JF635
Product Code. 30159530
  • £377.00 / 0.10mL
Description
SV40 Small T antigen Monoclonal antibody specifically detects SV40 Small T antigen in Virus samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SV40 Small T antigen
Monoclonal
Janelia Fluor 635
Mouse
Protein A purified
RUO
Virus
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
pab 280
50mM Sodium Borate
A purified Pab 419-SV40 Small T antigen
0.1 mL
Primary
Store at 4°C in the dark.
IgG1