missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV2C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21271-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.Specifications
| SV2C | |
| Polyclonal | |
| Unconjugated | |
| SV2C | |
| KIAA1054, SV2C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22987, 29643, 75209 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q496J9, Q69ZS6, Q9Z2I6 | |
| Sv2c | |
| SV2C Fusion Protein Ag15745 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction