missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SV2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561194
This item is not returnable.
View return policy
Description
Immunogen sequence: EYQGIPHPDD VKAKQAKMAP SRMDSLRGQT DLMAERLEDE EQLAHQYETI MDECGHGR Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 45%.
Probably plays a role in the control of regulated secretion in neural and endocrine cells.
Specifications
| SV2B | |
| Polyclonal | |
| Unconjugated | |
| Sv2b | |
| A830038F04Rik; AI850768; HsT19680; Kiaa0735; mKIAA0735; SV2B; Sv2bb; synaptic vesicle glycoprotein 2 b; synaptic vesicle glycoprotein 2b; Synaptic vesicle protein 2B; synaptic vesicle protein 2B homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L1I2 | |
| Sv2b | |
| Recombinant protein corresponding to Human SV2B. Recombinant protein control fragment (Product #RP-98176). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction