missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SV2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552476
This item is not returnable.
View return policy
Description
Immunogen sequence: SDGYYRGEGT QDEEEGGASS DATEGHDEDD EIYEGEYQGI PRAESGGKGE RMADGAPLAG VRGGLSDGEG PPGGRGEAQR RKEREELAQQ YEAILRECGH GRFQW Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This protein plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.It is identified as the brain binding-site for the antiepileptic drug levetiracetam/lev.
Specifications
| SV2A | |
| Polyclonal | |
| Unconjugated | |
| SV2A | |
| AI746429; calcium regulator SV2A; Kiaa0736; mKIAA0736; PSEC0174; SV2; SV2A; synaptic vesicle glycoprotein 2 a; synaptic vesicle glycoprotein 2a; Synaptic vesicle protein 2; synaptic vesicle protein 2A; synaptic vesicle transmembrane transporter (SV2) RNA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L0J3 | |
| SV2A | |
| Recombinant protein corresponding to Human SV2A. Recombinant protein control fragment (Product #RP-89907). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction