missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SV2A Monoclonal Antibody (K70011_14F1), Invitrogen™

Product Code. 30195650
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30195650 200 μL 200µg
30207386 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30195650 Supplier Invitrogen™ Supplier No. MA556040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: FIRGAKDIAK EVKKHAAKKV VKGLDRVQDE YSRRSYSRFE EEDDDDDFPA PSDGYYRGEG TQDEEEGGAS SDATEGHDED DEIYEGEYQG IPRAESGGKG ERMADGAPLA GVRGGLSDGE GPPGGRGEAQ RRKEREELAQ QYEAILRECG

This protein plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.It is identified as the brain binding-site for the antiepileptic drug levetiracetam/lev.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV2A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K70011_14F1
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SV2A
Gene Accession No. Q02563, Q7L0J3
Gene Alias AI746429; calcium regulator SV2A; Kiaa0736; mKIAA0736; PSEC0174; SV2; SV2A; synaptic vesicle glycoprotein 2 a; synaptic vesicle glycoprotein 2a; Synaptic vesicle protein 2; synaptic vesicle protein 2A; synaptic vesicle transmembrane transporter (SV2) RNA
Gene Symbols SV2A
Host Species Mouse
Immunogen Recombinant human SV2A (immunogen range is 11-160).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117559, 9900
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.