missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUZ12 Recombinant Rabbit Monoclonal Antibody (1B10)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA549827
This item is not returnable.
View return policy
Description
SUZ12 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA, IP
SUZ12 (polycomb protein SUZ12) is a zinc finger gene and has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. SUZ12 is also a component of the PRC2/EED-EZH2 complex which methylates Lys-9 (H3K9me) and Lys-27 (H3K27me) of histone H3 leading to transcriptional repression of the affected target gene. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
Specifications
| SUZ12 | |
| Recombinant Monoclonal | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15022 | |
| SUZ12 | |
| A synthesized peptide derived from human SUZ12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1B10 | |
| Unconjugated | |
| SUZ12 | |
| 2610028O16Rik; AI195385; AU016842; AW536442; chET 9 protein; CHET9; chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; Kiaa0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog; suppressor of zeste 12 homolog (Drosophila); suppressor of zeste 12 protein homolog; Suz12; SUZ12 polycomb repressive complex 2 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23512 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction