missing translation for 'onlineSavingsMsg'
Learn More

SUZ12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18331054 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331054 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331054 Supplier Novus Biologicals Supplier No. NBP310944100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SUZ12 Polyclonal specifically detects SUZ12 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUZ12
Applications ChIP Assay, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias ChET 9 protein, CHET9polycomb protein SUZ12, Chromatin precipitated E2F target 9 protein, JJAZ1Joined to JAZF1 protein, KIAA0160Suppressor of zeste 12 protein homolog, suppressor of zeste 12 homolog (Drosophila)
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human SUZ12 (NP_056170). Peptide sequence TGETNDKSTAPIAKPLATRNSESLHQENKPGSVKPTQTIAVKESLTTDLQ
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23512
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.