missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUZ12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20366-1-AP-150UL
This item is not returnable.
View return policy
Description
This zinc finger gene has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.Specifications
| SUZ12 | |
| Polyclonal | |
| Unconjugated | |
| SUZ12 | |
| ChET 9 protein, CHET9, JJAZ1, Joined to JAZF1 protein, KIAA0160, Polycomb protein SUZ12, SUZ12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23512, 52615, 688041 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15022, Q80U70 | |
| Suz12 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction